Supelco® Organic Reference Standard, Ginkgolide A
Catalog # 51863-10MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:15291-75-5
- Description:Ginkgolide A
- Seq:2275
- Storage conditions:–20 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00133365
- M-Clarity quality level:MQ100
- Molecular weight:408,40
Specifications
About this item
Caution: P302 + P352 - P305 + P351 + P338