Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, (+)-Pulegone
Supelco® Organic Reference Standard, (+)-Pulegone
Catalog # 82569-5ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (+)-Pulegone
Catalog # 82569-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    89-82-7
  • Description:
    (+)-Pulegone
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H351
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00063000
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (R)-2-Isopropylidene-5-methylcyclohexanone; (R)-p-Menth-4(8)-en-3-one
  • Molecular weight:
    152,23
  • Density:
    0,935 g/ml at 20 °C (lit.)
  • Flash point:
    82 °C - Closed cup
  • Boiling point:
    223 - 224 °C (lit.)

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P202 - P264 - P280 - P301 + P312 - P308 + P313 - P405