Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Disulfoton-sulfone
Supelco® PESTANAL® Organic Reference Standard, Disulfoton-sulfone
Catalog # 45871-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Disulfoton-sulfone
Catalog # 45871-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    2497-06-5
  • Description:
    Disulfoton-sulfone
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H300
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00155401
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    306,40

Specifications

About this item

  • GHS signal: GHS06
Caution: P264 - P270 - P301 + P310 - P405 - P501