Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Taxifolin
Supelco® Organic Reference Standard, Taxifolin
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Taxifolin
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    480-18-2
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00006845
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2R,3R)-3,3′,4′,5,7-Pentahydroxyflavanone; (2R,3R)-Dihydroquercetin
  • Molecular weight:
    304,25

Specifications

Related Information

Frequently Bought Together