Sigma-Aldrich® Needles, double tipped
Supplier: Merck
Certificates
About this item
Double tipped transfer needles with a non coring point on both ends.
Frequently Bought Together
Double tipped transfer needles with a non coring point on both ends.