Order Entry
Slovakia
ContactUsLinkComponent
N,N-Dimethylformamide for headspace gas chromatography
N,N-Dimethylformamide for headspace gas chromatography
Catalog # SCARDI10741000
Supplier:  SCHARLAB
CAS Number:  
undefined
N,N-Dimethylformamide for headspace gas chromatography
Catalog # SCARDI10741000
Supplier:  SCHARLAB
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284