Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Acetochlor
Supelco® PESTANAL® Organic Reference Standard, Acetochlor
Catalog # 33379-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Acetochlor
Catalog # 33379-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    34256-82-1
  • Description:
    Acetochlor
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H317 - H332 - H335 - H351 - H361f - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00143534
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Chloro-N-ethoxymethyl-N-(2′-ethyl-6′-methylphenyl) acetamide
  • Molecular weight:
    269,77

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P202 - P273 - P280 - P302 + P352 - P304 + P340 + P312 - P308 + P313