CHROMABOND® Vacuum Manifolds
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® Vacuum manifolds are designed for reproducible SPE applications.
- For simultaneous preparation of up to 12, 16 or 24 samples
- Available replacement parts for vacuum manifolds and accessories for special applications
Delivery: The vacuum manifold complete consists of glass cabinet with lid and lid gasket, removable needles on lower side of lid, vacuum gauge, control valve, valves and caps, variable rack, and PP tank (only for MANA730150).
Frequently Bought Together