Order Entry
Slovakia
ContactUsLinkComponent
 
Diethylene glycol ≥99.0%, Reagent Grade
Catalog # 0194-1L
Supplier:  VWR Chemicals
CAS Number:  
undefined
Diethylene glycol ≥99.0%, Reagent Grade
Catalog # 0194-1L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Color (APHA):
    ≤10
  • Purity:
    ≥99%
  • Refractive Index:
    1.445 – 1.450

Specification Test Results

About this item

 :
 :106.12 g/mol
Merck Index:13,03146
Boiling Pt:245 °C (1013 hPa)
Melting Pt:–10 °C
Density:1.118 g/cm³ (20 °C)
Flash Pt:123 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-872-2
 :GHS07
CAS Number:111-46-6
Formula:C₄H₁₀O₃
MDL Number:MFCD00002882