Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Clopyralid
  36758-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Clopyralid
  36758-250MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1702-17-6
  • Description:
    Clopyralid
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078655
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,6-Dichloro-pyridine-2-carboxylic acid
  • Molecular weight:
    192,00
  • Flash point:
    >100 °C - closed cup

 

 

  • GHS signal: GHS05
 : P280 - P305 + P351 + P338