Supelco® PESTANAL® Organic Reference Standard, Clopyralid (2-Hydroxyethyl)Ammonium
Catalog # 36529-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:57754-85-5
- Description:Clopyralid (2-hydroxyethyl)ammonium
- Storage conditions:2...30 °C
- Hazard class description:H318
- Shipping conditions:STD
- MDL No.:MFCD00777778
- M-Clarity quality level:MQ100
- Synonym(s):3,6-Dichloropyridine-2-carboxylic acid ethanolammonium salt
- Molecular weight:253,08
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS05
Caution: P280 - P305 + P351 + P338