3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate, Sigma-Aldrich®
Catalog # SIAL446130-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:422.82 g/mol
:M-Clarity™ quality level = MQ100
EINECS:241-165-0
CAS Number:17096-07-0
Formula:C₁₆H₃₈O₅Si₄
MDL Number:MFCD00053871