Estriol
Catalog # EHERC13213200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
EINECS:200-022-2
CAS Number:50-27-1
Formula:C₁₈H₂₄O₃
MDL Number:MFCD00003691