Pesticide reference standard, Chlordane 1000 µg/ml in isooctane, Supelco®
Catalog # 48065-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- CAS No.:57-74-9
- Description:Chlordane 1,000 µg/ml in isooctane
- M-Clarity quality level:MQ100
- Formula:C₁₀H₆Cl₈
- Description:Chlordane