Order Entry
Slovakia
ContactUsLinkComponent
Reinecke's salt, BAKER ANALYZED® ACS, J.T.Baker®
Reinecke's salt, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1388.0025
Supplier:  Avantor
CAS Number:  
Reinecke's salt, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1388.0025
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay:
    min. 93.0 %
  • Insoluble in Dilute HCl:
    max. 0.05 %
  • Sensitivity:
    passes test

Specification Test Results

About this item

 :
EINECS:237-003-3
CAS Number:13573-16-5
Formula:C₄H₁₀CrN₇S₄
MDL Number:MFCD00151654