Order Entry
Slovakia
ContactUsLinkComponent
sf540x390.png
Merck 3S reagents for titration with Metrohm OMNIS
3S is an intelligent packaging solution protected with an originality seal. There is an automatic data transfer between the cap and the Metrohm OMNIS titrator using RFID (radio-frequency identification).
Safe Safe chemical handling: minimized risk of contamination through contact-free reagent exchange
Smart Smart connection of chemical consumables: communication between the 3S cap and instrument software via RFID with a visual status indicator on the system
Secure Secure information transfer: Direct RFID-based transfer of reagent data to instrument software for elimination of human error. How to use
  • Remove tamper evident ring and push down to open
  • Connect with a single hand movement to the Omnis liquid adapter
  • Reagent data is automatically transferred to the method