Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FERRIC CHLORIDE TS, USP TEST SOLUTION 1 * 100 mL
Catalog # SPCMF-166-100ML
Supplier:
Spectrum Chemical
New Product
Expand image
FERRIC CHLORIDE TS, USP TEST SOLUTION 1 * 100 mL
Catalog # SPCMF-166-100ML
Supplier:
Spectrum Chemical
New Product
Frequently Bought Together
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW