25f-lmarkg-ptfe-5.1/0.72lc 25ul syr (lmk.2620517c-6p) pk6 1 * 6 items
Catalog # SGEAKLMK2620517C6P
Supplier: Trajan Scientific and Medical
New Product
25f-lmarkg-ptfe-5.1/0.72lc 25ul syr (lmk.2620517c-6p) pk6 1 * 6 items
Catalog # SGEAKLMK2620517C6P
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Collecting trays, b.safe
Supplier: Bohlender
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Uniguard™ direct connection guard cartridge holders for Accucore™ columns
Supplier: Thermo Fisher Scientific
Vial racks
Supplier: MACHEREY-NAGEL
Syringeless filters, Mini-UniPrep™, Whatman™
Supplier: Whatman products (Cytiva)
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Crimp caps with 11 mm septa
Supplier: Agilent
HPLC columns, Hypercarb™
Supplier: Thermo Fisher Scientific
Capillaries for LC systems
Supplier: Agilent
Immobilised polysaccharide columns
Supplier: CHIRAL TECHNOLOGIES
Closures for 13 mm screw neck vials, Chromacol™
Supplier: CHROMACOL
O-RING SCREW BEAD ADJUSTER NITRILE 1 * 1 items
Supplier: VARIAN
HPLC columns, ZORBAX® 300 Å StableBond®
Supplier: Agilent