5 components: Ca 2000mg/l Mg 250mg/l Na 10000mg/l S 2500mg/l Si 250mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM122355L1
Supplier: CUSTOM MADE CHEMICALS LAB