ContactUsLinkComponent
Muffle Furnace L9/13/C550 Tmax1300 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3229
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/13/C550 Tmax1300 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3229
Supplier:  NABERTHERM