ContactUsLinkComponent
Muffle Furnace L40/12/B510 Tmax1200 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3057
Supplier:  NABERTHERM
New Product
Muffle Furnace L40/12/B510 Tmax1200 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3057
Supplier:  NABERTHERM