ContactUsLinkComponent
Muffle Furnace L24/12/B510 Tmax1200 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2964
Supplier:  NABERTHERM
New Product
Muffle Furnace L24/12/B510 Tmax1200 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2964
Supplier:  NABERTHERM