ContactUsLinkComponent
Muffle Furnace L3/12/C550 Tmax1200 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2592
Supplier:  NABERTHERM
New Product
Muffle Furnace L3/12/C550 Tmax1200 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-2592
Supplier:  NABERTHERM