ContactUsLinkComponent
WEDO® SuperStep plastic kick stool, blue (RAL 5002) made of unbreakable, lightfast polypropylene with treads made of TPE. The slip resistance of the treads is improved by the joint processing of the 2 components so that slipping is practically imposs 1 * 1 items
WEDO® Super Step Kick Stools
Catalog # WEDO212303
Supplier:  WEDO WERNER DORSCH
New Product
undefined
WEDO® Super Step Kick Stools
Catalog # WEDO212303
Supplier:  WEDO WERNER DORSCH
New Product
Specifications
  • Colour:
    Blue
  • Height:
    428 mm, unloaded; 415 mm loaded
  • Seq:
    2
  • Ø top/Ø bottom:
    292 mm/424 mm
Specifications
About this item
The WEDO® Super Step castor stool is made of unbreakable, lightfast polypropylene with treads made of TPE.
  • Unbreakable
    Rollable
    Non-slip treads
The slip resistance of the treads is improved by the joint processing of the two components so that slipping is practically impossible.
The treads cannot be removed. As both plastics come from the same family, the Super Step body can still be 100% recycled and is assembled by simply plugging the upper and lower parts together. No tools are required.
The three replaceable castors mounted on steel springs are pre-assembled. Under load, the Super Step® stands firmly on its integrated rubber support ring.
Max. load capacity (tested and approved): 150 kg.
Certifications:
TÜV and GS tested in accordance with EN 14183-F.
Ordering information:
Ten years guarantee on the plastic body.
Frequently Bought Together