ContactUsLinkComponent
Eppendorf Conical Tubes 50 mL,sterile, pyrogen, DNase, RNase, and DNA free, 480 pcs., 24 packs of 20 Tubes each 1 * 480 items
Catalog # EPPE0030122.569
Supplier:  EPPENDORF
New Product
Eppendorf Conical Tubes 50 mL,sterile, pyrogen, DNase, RNase, and DNA free, 480 pcs., 24 packs of 20 Tubes each 1 * 480 items
Catalog # EPPE0030122.569
Supplier:  EPPENDORF