ContactUsLinkComponent
,,17 components: Cd 1mg/l Co 1mg/l Cr 1mg/l Cu 1mg/l Mn 1mg/l Ni 1mg/l Zn 1mg/l S 1mg/l Hg 1mg/l Pb 1mg/l V 1mg/l Sn 1mg/l As 1mg/l Se 1mg/l Sb 1mg/l Tl 1mg/l Sc 0.5mg/l in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,, ,,Co 1 * 100 mL
Catalog # CMPLRM110738L1
New Product
,,17 components: Cd 1mg/l Co 1mg/l Cr 1mg/l Cu 1mg/l Mn 1mg/l Ni 1mg/l Zn 1mg/l S 1mg/l Hg 1mg/l Pb 1mg/l V 1mg/l Sn 1mg/l As 1mg/l Se 1mg/l Sb 1mg/l Tl 1mg/l Sc 0.5mg/l in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,, ,,Co 1 * 100 mL
Catalog # CMPLRM110738L1