Orders LinkContactUsLinkComponent
TOC STANDARD SOLUTION 25 MG/ - 1L 1 * 1.000 mL
TOC STANDARD SOLUTION 25 MG/ - 1L 1 * 1.000 mL
Catalog # ACSDTOC25.1L
New Product
Supplier:   CPACHEM
TOC STANDARD SOLUTION 25 MG/ - 1L 1 * 1.000 mL
Catalog # ACSDTOC25.1L
Supplier:  CPACHEM
New Product
TOC STANDARD SOLUTION 25 MG/ - 1L 1 * 1.000 mL
Price
280,00 €
Log in for availability
1 * 1.000 mL