RadEye G-10, Pocket–sized wide range survey meter: energy compensated GM-tube according H*(10): 45 keV – 1.3 MeV measuring range 0,05 µSv/h – 100 mSv/h high sensitivity: 1.7 cps per µSv/h 2 dose and dose rate alarm levels red label 1 * 1 items
Catalog # HERA4250676
Supplier: Thermo Scientific