ContactUsLinkComponent
Goat IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270384-2096
Supplier:  ANTIBODIES.COM
New Product
Goat IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Catalog # ANTIA270384-2096
Supplier:  ANTIBODIES.COM