Human alpha 1a Adrenergic Receptor/ADRA1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human alpha 1a Adrenergic Receptor/ADRA1A in tissue homogenates, cell lysates or oth 1 * 96 Tests
Catalog # ANTIA2320-96
Supplier: ANTIBODIES.COM