ContactUsLinkComponent
Mouse TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 1.920 Tests
Catalog # ANTIA270530-2096
Supplier:  ANTIBODIES.COM
New Product
Mouse TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 1.920 Tests
Catalog # ANTIA270530-2096
Supplier:  ANTIBODIES.COM