ContactUsLinkComponent
Human Phospholipase C gamma 1/PLC-gamma-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Phospholipase C gamma 1/PLC-gamma-1 in tissue homogenates, cell lysates or oth 1 * 96 Tests
Catalog # ANTIA2654-96
Supplier:  ANTIBODIES.COM
New Product
Human Phospholipase C gamma 1/PLC-gamma-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Phospholipase C gamma 1/PLC-gamma-1 in tissue homogenates, cell lysates or oth 1 * 96 Tests
Catalog # ANTIA2654-96
Supplier:  ANTIBODIES.COM