ContactUsLinkComponent
Bovine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 576 Tests
Catalog # ANTIA270374-696
Supplier:  ANTIBODIES.COM
New Product
Bovine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 576 Tests
Catalog # ANTIA270374-696
Supplier:  ANTIBODIES.COM