Monkey IL-13 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant human IL-13 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Catalog # ANTIA270426-2096
Supplier: Antibodies.com