Human GM-CSF Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human GM-CSF ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Catalog # ANTIA270475-2096
Supplier: Antibodies.com