ContactUsLinkComponent
Mouse NF-kB p65 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse NF-kB p65 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA2082-96
Supplier:  ANTIBODIES.COM
New Product
Mouse NF-kB p65 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse NF-kB p65 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA2082-96
Supplier:  ANTIBODIES.COM