Mouse CGRP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinatio n of mouse CGRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluid 1 * 96 Tests
Catalog # ANTIA1728-96
Supplier: ANTIBODIES.COM