ContactUsLinkComponent
Mouse IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant mouse IL-17A ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Catalog # ANTIA270523-2096
Supplier:  Antibodies.com
New Product
Mouse IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant mouse IL-17A ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Catalog # ANTIA270523-2096
Supplier:  Antibodies.com