ContactUsLinkComponent
Human IgE Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, native human IgE ELISA standard, and standard reconstitution buffer. This matched antibody pair ki 1 * 576 Tests
Catalog # ANTIA270480-696
Supplier:  ANTIBODIES.COM
New Product
Human IgE Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, native human IgE ELISA standard, and standard reconstitution buffer. This matched antibody pair ki 1 * 576 Tests
Catalog # ANTIA270480-696
Supplier:  ANTIBODIES.COM