ContactUsLinkComponent
Mouse IL-12 p70 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse IL-12 p70 ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270522-696
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-12 p70 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse IL-12 p70 ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270522-696
Supplier:  ANTIBODIES.COM