Monkey beta Crosslaps/beta CTx ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of monkey beta Crosslaps/beta CTx in serum, plasma, cell culture supernatant, cell or tissue lysat 1 * 96 Tests
Catalog # ANTIA326974-96
Supplier: ANTIBODIES.COM