SARS-CoV-2 Spike Glycoprotein S2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of SARS-CoV-2 Spike Glycoprotein S2 in serum, plasma, cell culture supernatant, cell or tissue l 1 * 96 Tests
Catalog # ANTIA326870-96
Supplier: ANTIBODIES.COM