ContactUsLinkComponent
Mouse NFkB p100/NFKB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse NFkB p100/NFKB2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2859-96
Supplier:  ANTIBODIES.COM
New Product
Mouse NFkB p100/NFKB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse NFkB p100/NFKB2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2859-96
Supplier:  ANTIBODIES.COM