ContactUsLinkComponent
Bovine IL-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine IL-10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 48 Tests
Catalog # ANTIA2870-48
Supplier:  ANTIBODIES.COM
New Product
Bovine IL-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine IL-10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 48 Tests
Catalog # ANTIA2870-48
Supplier:  ANTIBODIES.COM