ContactUsLinkComponent
Mouse High Density Lipoprotein Cholesterol ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vi tro quantitative determination of mouse High Density Lipoprotein Cholesterol in serum, plasma, cell culture supernata 1 * 96 Tests
Catalog # ANTIA326613-96
Supplier:  ANTIBODIES.COM
New Product
Mouse High Density Lipoprotein Cholesterol ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vi tro quantitative determination of mouse High Density Lipoprotein Cholesterol in serum, plasma, cell culture supernata 1 * 96 Tests
Catalog # ANTIA326613-96
Supplier:  ANTIBODIES.COM