ContactUsLinkComponent
Mouse IL-12 p40 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse IL-12 p40 in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of samp 1 * 96 Tests
Catalog # ANTIA323227-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-12 p40 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse IL-12 p40 in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of samp 1 * 96 Tests
Catalog # ANTIA323227-96
Supplier:  ANTIBODIES.COM