ContactUsLinkComponent
Mouse Anti-SARS-CoV-2 Spike RBD IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-SARS-CoV-2 Spike RBD IgG in serum, plasma, cell culture supernatant, cell or tissue lysa 1 * 96 Tests
Catalog # ANTIA326399-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Anti-SARS-CoV-2 Spike RBD IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-SARS-CoV-2 Spike RBD IgG in serum, plasma, cell culture supernatant, cell or tissue lysa 1 * 96 Tests
Catalog # ANTIA326399-96
Supplier:  ANTIBODIES.COM