ContactUsLinkComponent
Beta Lactam + Tetracycline + Streptomycin + Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for th e qualitative determination of Beta Lactam + Tetracycline + Streptomycin + Chloramphenicol in raw commingled cow&#014 1 * 96 Tests
  ANTIA327185-96
New Product
Beta Lactam + Tetracycline + Streptomycin + Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for th e qualitative determination of Beta Lactam + Tetracycline + Streptomycin + Chloramphenicol in raw commingled cow&#014 1 * 96 Tests
  ANTIA327185-96