Dipyrone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Dipyrone in raw comm ingled cow’ s milk. 1 * 96 Tests
Catalog # ANTIA327200-96
Supplier: ANTIBODIES.COM