Aflatoxin B1 Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Aflatoxin B1 in feed, cereals, oil sample. 1 * 10 Tests
Catalog # ANTIA327177-10
Supplier: ANTIBODIES.COM